missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Histone H1.
Brand: Abnova PAB24849.100uL
This item is not returnable.
View return policy
Specifications
| Histone H1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| Histone H1 polyclonal antibody detects endogenous levels of protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of Histone H1. | |
| In 1X PBS, pH 7.2 (0.05% sodium azide) | |
| A synthetic peptide corresponding to Histone H1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction