missing translation for 'onlineSavingsMsg'
Learn More
Learn More
His2Av, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phsphopeptide of His2Av.
Brand: Abnova PAB9920.100ug
This item is not returnable.
View return policy
Specifications
| His2Av | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phsphopeptide of His2Av. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| CG5499/H2A.X/H2A.Z/H2AV/H2AX/H2AZ/H2AvD/His2AvD/l(3)05146/l(3)810/l(3)97Dd/l(3)L1602 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Reactivity occurs against Drosophila H2AvD pS137 protein and This antibody is specific to the phosphorylated form of the protein. Reactivity with non-phosphorylated Drosophila H2AvD is minimal by ELISA. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:4000-1:16000) Western Blot (1:400-1:1600) The optimal working dilution should be determined by the end user. | |
| His2Av | |
| His2Av | |
| Synthetic phosphopeptide corresponding to residues surrounding S137 of drosophila His2Av. | |
| RUO | |
| 43229 | |
| Fruitfly | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction