missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEXIM1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HEXIM1.
Brand: Abnova PAB18762.100ug
This item is not returnable.
View return policy
Description
Expression of this gene is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells. This gene has no introns. [provided by RefSeq
Specifications
| HEXIM1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HEXIM1. | |
| In buffer containing 0.02% sodium azide | |
| CLP1/EDG1/FLJ13562/HIS1/MAQ1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:20000-1:80000) Western Blot (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| HEXIM1 | |
| HEXIM1 | |
| A synthetic peptide corresponding to 15 amino acids at internal region of human HEXIM1. | |
| RUO | |
| 10614 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction