Learn More
HES6, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HES6.
Brand: Abnova PAB24925.100uL
Description
This gene encodes a member of a subfamily of basic helix-loop-helix transcription repressors that have homology to the Drosophila enhancer of split genes. Members of this gene family regulate cell differentiation in numerous cell types. The protein encoded by this gene functions as a cofactor, interacting with other transcription factors through a tetrapeptide domain in its C-terminus. Alternatively spliced transcript variants encoding different isoforms have been described
Specifications
| HES6 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| HES6 | |
| HES6 | |
| A synthetic peptide corresponding to HES6. | |
| 100 μL | |
| Primary | |
| HES6 polyclonal antibody detects endogenous levels of HES6 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of HES6. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| HES-6/bHLHb41/bHLHc23 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55502 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only