Learn More
HDAC1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HDAC1.
Brand: Abnova PAB0180.100ug
Description
Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. The protein encoded by this gene belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, it deacetylates p53 and modulates its effect on cell growth and apoptosis. [provided by RefSeq
Specifications
| HDAC1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HDAC1. | |
| In PBS (0.02% sodium azide) | |
| DKFZp686H12203/GON-10/HD1/RPD3/RPD3L1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| HDAC1 | |
| HDAC1 | |
| A mixture of synthetic peptides corresponding to amino acids 1-5, 433-448 and 467-482 of human HDAC1. | |
| RUO | |
| 3065 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only