Learn More
HCFC2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant HCFC2.
Brand: Abnova PAB20343.100uL
Description
This gene encodes one of two proteins which interact with VP16, a herpes simplex virus protein that initiates virus infection. Both the encoded protein and the original Herpes host cell factor interact with VP16 through a beta-propeller domain. The original Herpes host cell factor, however, is effective at initiating viral infection while the encoded protein is not. Transcripts of varying length due to alternative polyadenylation signals have been described. [provided by RefSeq
Sequence: APSQVQLIKATTNSFHVKWDEVSTVEGYLLQLSTDLPYQAASSDSSAAPNMQGVRMDPHRQGSNNIVPNSINDTINSTKTEQPATKETSMKNKPDFKALTDSNAILYPSLASNASNHNSHVVDMLRKNEGPHTSANVGVSpecifications
| HCFC2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant HCFC2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ94012/HCF-2/HCF2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 29915 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| HCFC2 | |
| HCFC2 | |
| Recombinant protein corresponding to amino acids of human HCFC2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only