Learn More
HAT1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of HAT1.
Brand: Abnova PAB2245.400uL
Description
The protein encoded by this gene is a type B histone acetyltransferase (HAT) that is involved in the rapid acetylation of newly synthesized cytoplasmic histones, which are in turn imported into the nucleus for de novo deposition onto nascent DNA chains. Histone acetylation, particularly of histone H4, plays an important role in replication-dependent chromatin assembly. Specifically, this HAT can acetylate soluble but not nucleosomal histone H4 at lysines 5 and 12, and to a lesser degree, histone H2A at lysine 5. Alternatively spliced transcript variants have been identified for this gene. [provided by RefSeq
Specifications
| HAT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of HAT1. | |
| In PBS (0.09% sodium azide) | |
| KAT1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 8520 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| HAT1 | |
| HAT1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human HAT1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only