missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H3K79me1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of H3K79me1.
Brand: Abnova PAB14056.100uL
This item is not returnable.
View return policy
Specifications
| H3K79me1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of H3K79me1. | |
| In serum (0.05% sodium azide) | |
| A synthetic peptide (conjugated with KLH) corresponding to region of histone H3 containing the monomethylated lysine 79 (H3K79me1). | |
| RUO | |
| Human | |
| Liquid |
| ChIP Assay, Dot Blot, ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:500-1:1000) Dot Blot (1:100000) Western Blot (1:1000) ChIP (5-10 ul/ChIP) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction