Learn More
GUCY1B3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GUCY1B3.
Brand: Abnova PAB21333.100uL
Description
Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha subunit and a beta subunit, typically GUCY1B3, catalyzes conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide (NO) and nitrovasodilator drugs (Zabel et al., 1998 [PubMed 9742212]).[supplied by OMIM
Sequence: HALELLVIRNYGPEVWEDIKKEAQLDEEGQFLVRIIYDDSKTYDLVAAASKVLNLNAGEILQMFGKMFFVFCQESGYDTILRVLGSNVREFLQNLDALHDHLATIYPGMRAPSFRCTDAEKGKGLILHYYSEREGLQDIVIGIISpecifications
| GUCY1B3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GUCY1B3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q02153 | |
| GUCY1B3 | |
| Recombinant protein corresponding to amino acids of human GUCY1B3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| GUCY1B3 | |
| GC-S-beta-1/GC-SB3/GUC1B3/GUCB3/GUCSB3/GUCY1B1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2983 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only