Learn More
GUCY1B3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GUCY1B3.
Brand: Abnova PAB4059.400uL
Description
Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha subunit and a beta subunit, typically GUCY1B3, catalyzes conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide (NO) and nitrovasodilator drugs (Zabel et al., 1998 [PubMed 9742212]).[supplied by OMIM
Specifications
| GUCY1B3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GUCY1B3. | |
| In PBS (0.09% sodium azide) | |
| NP_000848 | |
| GUCY1B3 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GUCY1B3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| GUCY1B3 | |
| GC-S-beta-1/GC-SB3/GUC1B3/GUCB3/GUCSB3/GUCY1B1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 2983 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only