Learn More
GUCY1A2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GUCY1A2.
Brand: Abnova PAB4064.400uL
Description
Soluble guanylyl (or guanylate) cyclases are heterodimeric enzymes consisting of an alpha subunit, such as alpha-2 (GUCY1A2), and a beta subunit, typically beta-1 (GUCY1B3; MIM 139397), which are activated by nitric oxide (NO) and which catalyze conversion of GTP to 3-prime, 5-prime-cyclic GMP and pyrophosphate (Harteneck et al., 1991 [PubMed 1683630]).[supplied by OMIM
Specifications
| GUCY1A2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GUCY1A2. | |
| In PBS (0.09% sodium azide) | |
| NP_000846 | |
| GUCY1A2 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GUCY1A2. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:50-200) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| GUCY1A2 | |
| GC-SA2/GUC1A2 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 2977 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only