Learn More
GTPBP10, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GTPBP10.
Brand: Abnova PAB21354.100uL
Description
Small G proteins, such as GTPBP10, act as molecular switches that play crucial roles in the regulation of fundamental cellular processes such as protein synthesis, nuclear transport, membrane trafficking, and signal transduction (Hirano et al., 2006 [PubMed 17054726]).[supplied by OMIM
Sequence: ALKGSKGKDCEIPVPVGISVTDENGKIIGELNKENDRILVAQGGLGGKLLTNFLPLKGQKRIIHLDLKLIADVGLVGFPNAGKSSLLSCVSHAKPAIADYAFSpecifications
| GTPBP10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GTPBP10. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| A4D1E9 | |
| GTPBP10 | |
| Recombinant protein corresponding to amino acids of human GTPBP10. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| GTPBP10 | |
| DKFZp686A10121/FLJ38242/MGC104191/ObgH2/UG0751c10 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 85865 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only