Learn More
GSK3B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GSK3B.
Brand: Abnova PAB3336.400uL
Description
The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Specifications
| GSK3B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GSK3B. | |
| In PBS (0.09% sodium azide) | |
| P49841 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 2932 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| GSK3B | |
| GSK3B | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human GSK3B. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only