Learn More
GSK3B (phospho Y216), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GSK3B.
Brand: Abnova PAB12624.100ug
Description
The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Sequence: NVSYISpecifications
| GSK3B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of GSK3B. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| GSK3B | |
| Synthetic phosphopeptide corresponding to residues surrounding Y216 of human GSK3B. | |
| RUO | |
| 2932 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| GSK3B | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes GSK3B (pY216) with a phosphorylated site at Tyrosine 216. It does not cross-react with non-phosphospecific peptide. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only