Learn More
GRK1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRK1.
Brand: Abnova PAB4515.400uL
Description
This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates rhodopsin and initiates its deactivation. Defects in GRK1 are known to cause Oguchi disease 2 (also known as stationary night blindness Oguchi type-2). [provided by RefSeq
Specifications
| GRK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GRK1. | |
| In PBS (0.09% sodium azide) | |
| Q15835 | |
| GRK1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GRK1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| GRK1 | |
| GPRK1/RHOK/RK | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 6011 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only