missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIP1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GRIP1.
Brand: Abnova PAB27234.100ug
This item is not returnable.
View return policy
Specifications
| GRIP1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) Immunofluorescence (20 ug/mL) The optimal working dilution should be determined by the end user. | |
| GRIP1 | |
| GRIP | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 23426 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of GRIP1. | |
| In PBS (0.02% sodium azide) | |
| NP_066973 | |
| GRIP1 | |
| A synthetic peptide corresponding to 17 amino acids at C-terminus of human GRIP1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction