missing translation for 'onlineSavingsMsg'
Learn More

GRINA Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant GRINA.

Brand:  Abnova PAB23092.100uL

Product Code. 16125680

  • £535.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: PYAQPPYPGAPYPQPPFQPSPYGQPGYPHGPSPYPQGGYPQGPY
Specifications

Specifications

GRINA
Polyclonal
Rabbit polyclonal antibody raised against recombinant GRINA.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Q7Z429
GRINA
Recombinant protein corresponding to amino acids of human GRINA.
100 μL
Primary
Human
Liquid
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
GRINA
HNRGW/MGC99687/NMDARA1/TMBIM3
Rabbit
Antigen affinity purification
RUO
2907
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only