missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grin2b Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Grin2b.
Brand: Abnova PAB9700.10ug
This item is not returnable.
View return policy
Specifications
| Grin2b | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Grin2b. | |
| Lyophilized from 5mM ammonium bicarbonate | |
| Q00960 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 24410 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Frozen sections) (1:1000-2000) Immunoprecipitation (3 ul/200 ug lysate) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Grin2b | |
| Grin2b | |
| Recombinant protein corresponding to C-terminus rat Grin2b. | |
| 10 μg | |
| Primary | |
| Recognizes human, mouse and rat forms of the NR2B subunits of NMDAR. | |
| Store at -20°C on dry atmosphere. After reconstitution with PBS store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction