missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grin2b Rabbit anti-Bovine, Chicken, Dog, Human, Mouse, Primates, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Grin2b.
Brand: Abnova PAB9636.100uL
This item is not returnable.
View return policy
Specifications
| Grin2b | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Grin2b. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| Q00960 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 24410 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Grin2b | |
| Grin2b | |
| A synthetic peptide corresponding to N-terminus rat Grin2b. | |
| 100 μL | |
| Primary | |
| Bovine, Canine, Chicken, Human, Mouse, Primate, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction