missing translation for 'onlineSavingsMsg'
Learn More

GRIA2 (phospho S880), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic phosphopeptide of GRIA2.

Brand:  Abnova PAB25871.100ug

Product Code. 16096047

  • £349.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to a family of glutamate receptors that are sensitive to alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate (AMPA), and function as ligand-activated cation channels. These channels are assembled from 4 related subunits, GRIA1-4. The subunit encoded by this gene (GRIA2) is subject to RNA editing (CAG->CGG; Q->R) within the second transmembrane domain, which is thought to render the channel impermeable to Ca(2+). Human and animal studies suggest that pre-mRNA editing is essential for brain function, and defective GRIA2 RNA editing at the Q/R site may be relevant to amyotrophic lateral sclerosis (ALS) etiology. Alternative splicing, resulting in transcript variants encoding different isoforms, (including the flip and flop isoforms that vary in their signal transduction properties), has been noted for this gene. [provided by RefSeq]

Sequence: I-E-Sp-V-K
Specifications

Specifications

GRIA2
Polyclonal
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
GRIA2
GLUR2/GLURB/GluR-K2/HBGR2
Rabbit
Affinity chromatography
RUO
2891
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Western Blot
1 mg/mL
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GRIA2.
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
P42262
GRIA2
Synthetic phosphopeptide corresponding to residues surrounding S880 of human GRIA2.
100 μg
Primary
Human, Mouse, Rat
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only