Learn More
GPSN2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GPSN2.
Brand: Abnova PAB22340.100uL
Description
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2,3-enoyl-CoA to saturated acyl-CoA (Moon and Horton, 2003 [PubMed 12482854]).[supplied by OMIM
Sequence: TQMTIWAKGKHRSYLKEFRDYPPLRMPIISpecifications
| GPSN2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GPSN2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9NZ01 | |
| GPSN2 | |
| Recombinant protein corresponding to amino acids of human GPSN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| GPSN2 | |
| SC2/TER | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9524 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only