Learn More
GPSM2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GPSM2.
Brand: Abnova PAB20424.100uL
Description
Heterotrimeric G proteins transduce extracellular signals received by cell surface receptors into integrated cellular responses. GPSM2 belongs to a group of proteins that modulate activation of G proteins (Blumer et al., 2002 [PubMed 11832491]).[supplied by OMIM
Sequence: FQSNRMDDQRCCLQEKNCHTASTTTSSTPPKMMLKTSSVPVVSPNTDEFLDLLASSQSRRLDDQRASFSNLPGLRLTQNSQSVLSHLMTNDNKEADEDFFDSpecifications
| GPSM2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GPSM2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P81274 | |
| GPSM2 | |
| Recombinant protein corresponding to amino acids of human GPSM2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| GPSM2 | |
| LGN/Pins | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 29899 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only