Learn More
GPRC5C Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPRC5C.
Brand: Abnova PAB16352.50ug
Description
The protein encoded by this gene is a member of the type 3 G protein-coupled receptor family. Members of this superfamily are characterized by a signature 7-transmembrane domain motif. The specific function of this protein is unknown; however, this protein may mediate the cellular effects of retinoic acid on the G protein signal transduction cascade. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| GPRC5C | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPRC5C. | |
| In PBS (0.09% sodium azide) | |
| Q9NQ84 | |
| GPRC5C | |
| A synthetic peptide (conjugated with KLH) corresponding to human GPRC5C. | |
| 50 μg | |
| Primary | |
| C-terminal Cytoplasmic domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPRC5C | |
| MGC131820/RAIG-3/RAIG3 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 55890 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only