Learn More
GPRC5A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPRC5A.
Brand: Abnova PAB26484.50ug
Description
This gene encodes a member of the type 3 G protein-coupling receptor family, characterized by the signature 7-transmembrane domain motif. The encoded protein may be involved in interaction between retinoid acid and G protein signalling pathways. Retinoic acid plays a critical role in development, cellular growth, and differentiation. This gene may play a role in embryonic development and epithelial cell differentiation. [provided by RefSeq
Specifications
| GPRC5A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPRC5A. | |
| In PBS (0.09% sodium azide) | |
| Q8NFJ5 | |
| GPRC5A | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human GPRC5A. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (18-36 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPRC5A | |
| GPCR5A/RAI3/RAIG1 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 9052 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only