Learn More
GPR85 Rabbit anti-Bovine, Chicken, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Zebra fish, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR85.
Brand: Abnova PAB26481.50ug
Description
Members of the G protein-coupled receptor (GPCR) family, such as GPR85, have a similar structure characterized by 7 transmembrane domains. Activation of GPCRs by extracellular stimuli, such as neurotransmitters, hormones, or light, induces an intracellular signaling cascade mediated by heterotrimeric GTP-binding proteins, or G proteins (Matsumoto et al., 2000 [PubMed 10833454]).[supplied by OMIM
Specifications
| GPR85 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR85. | |
| In PBS (0.09% sodium azide) | |
| P60893 | |
| GPR85 | |
| A synthetic peptide corresponding to 20 amino acids at C-terminus of human GPR85. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except NLRC3 (40%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (9-17 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR85 | |
| SREB/SREB2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 54329 | |
| Bovine, Canine, Chicken, Equine, Fish, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only