Learn More
GPR55 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR55.
Brand: Abnova PAB26551.50ug
Description
Members of the G protein-coupled receptor (GPR) family, such as GPR55, play important roles in signal transduction from the external environment to the inside of the cell (Sawzdargo et al., 1999 [PubMed 9931487]).[supplied by OMIM
Specifications
| GPR55 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR55. | |
| In PBS (0.09% sodium azide) | |
| Q9Y2T6 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 9290 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR55 | |
| GPR55 | |
| A synthetic peptide corresponding to 20 amino acids at C-terminus of human GPR55. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only