Learn More
GPR183, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR183.
Brand: Abnova PAB26365.50ug
Additional Details : Weight : 0.02000kg
Description
This gene was identified by the up-regulation of its expression upon Epstein-Barr virus infection of primary B lymphocytes. This gene is predicted to encode a G protein-coupled receptor that is most closely related to the thrombin receptor. Expression of this gene was detected in B-lymphocyte cell lines and lymphoid tissues but not in T-lymphocyte cell lines or peripheral blood T lymphocytes. The function of this gene is unknown. [provided by RefSeq
Specifications
GPR183 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of GPR183. | |
In PBS (0.09% sodium azide) | |
P32249 | |
GPR183 | |
A synthetic peptide corresponding to 15 amino acids at C-terminus of human GPR183. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
GPR183 | |
EBI2 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
1880 | |
Human | |
Liquid |
For Research Use Only