Learn More
GPR182 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR182.
Brand: Abnova PAB5229.100ug
Description
Adrenomedullin is a potent vasodilator peptide that exerts major effects on cardiovascular function. This gene encodes a seven-transmembrane protein that belongs to the family 1 of G-protein coupled receptors. Studies of the rat counterpart suggest that the encoded protein may function as a receptor for adrenomedullin. [provided by RefSeq]
Specifications
| GPR182 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR182. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| 7TMR/ADMR/AM-R/AMR/G10D/MGC34399/gamrh/hrhAMR | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| GPR182 | |
| GPR182 | |
| A synthetic peptide corresponding to N-terminus of human GPR182. | |
| RUO | |
| 11318 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only