missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR160 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR160.
Brand: Abnova PAB16390.50ug
This item is not returnable.
View return policy
Specifications
| GPR160 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR160. | |
| In PBS (0.09% sodium azide) | |
| Q9UJ42 | |
| GPR160 | |
| A synthetic peptide (conjugated with KLH) corresponding to human GPR160. | |
| 50 μg | |
| Primary | |
| 3rd Cytoplasmic domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR160 | |
| GPCR1/GPCR150 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 26996 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction