missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR124 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR124.
Brand: Abnova PAB13308.100ug
This item is not returnable.
View return policy
Specifications
| GPR124 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR124. | |
| In PBS (0.02% sodium azide) | |
| Q96PE1 | |
| GPR124 | |
| A synthetic peptide corresponding to internal region 14 amino acids of human GPR124. | |
| RUO | |
| 25960 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR124 | |
| DKFZp434C211/DKFZp434J0911/FLJ14390/KIAA1531/TEM5 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction