Learn More
GPER Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GPER.
Brand: Abnova PAB22000.100uL
Description
This gene is a member of the G-protein coupled receptor 1 family and encodes a multi-pass membrane protein that localizes to the endoplasmic reticulum. The protein binds estrogen, resulting in intracellular calcium mobilization and synthesis of phosphatidylinositol 3,4,5-trisphosphate in the nucleus. This protein therefore plays a role in the rapid nongenomic signaling events widely observed following stimulation of cells and tissues with estrogen. Alternate transcriptional splice variants which encode the same protein have been characterized. [provided by RefSeq
Sequence: MDVTSQARGVGLEMYPGTAQPAAPNTTSPELNLSHPLLGTALANGTGELSEHQQYVIGLFLSSpecifications
| GPER | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GPER. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q99527 | |
| GPER | |
| Recombinant protein corresponding to amino acids of human GPER. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| GPER | |
| CEPR/CMKRL2/DRY12/FEG-1/GPCR-Br/GPR30/LERGU/LERGU2/LyGPR/MGC99678 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2852 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only