missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOPC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GOPC.
Brand: Abnova PAB21636.100uL
This item is not returnable.
View return policy
Description
PIST is a PDZ domain-containing Golgi protein. PDZ domains contain approximately 90 amino acids and bind the extreme C terminus of proteins in a sequence-specific manner.[supplied by OMIM
Sequence: IEFEVVYVAPEVDSDDENVEYEDESGHRYRLYLDELEGGGNPGASCKDTSGEIKVLQGFNKKAVTDTHENGDLGTASETPLDDGASKLDDLHTLYSpecifications
| GOPC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GOPC. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9HD26 | |
| GOPC | |
| Recombinant protein corresponding to amino acids of human GOPC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| GOPC | |
| CAL/FIG/GOPC1/PIST/dJ94G16.2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57120 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction