Learn More
GOLIM4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GOLIM4
Brand: Abnova PAB28477.100uL
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi-resident protein. It may process proteins synthesized in the rough endoplasmic reticulum and assist in the transport of protein cargo through the Golgi apparatus. [provided by RefSeq
Sequence: EHLEEEHDPSPEEQDREWKEQHEQREAANLLEGHARAEVYPSAKPMIKFQSPYEEQLEQQRLAVQQVEEAQQLREHQEALHQQRLQGHLLRQQEQQQQQVAREMALQRQAELEEGRPQHQEQLRQQAHYDAMDNDIVQGAEDSpecifications
| GOLIM4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GOLIM4 | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| F8W785 | |
| GOLIM4 | |
| Recombinant protein corresponding to amino acids of human GOLIM4 | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry(1:200-1:500) Immunofluorescence(1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| GOLIM4 | |
| GIMPC/GOLPH4/GPP130/P138 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 27333 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only