missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody, DyLight 680, Novus Biologicals™

Product Code. 18420487 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mL
Unit Size:
0.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420487 0.5 mL 0.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420487 Supplier Novus Biologicals Supplier No. NBP172726FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Golden Syrian Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Western Blot, ELISA, Immunohistochemistry, Dot Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Golden Syrian Hamster IgG whole molecule
Purification Method Immunogen affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Golden Syrian Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.