missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Golden Syrian Hamster IgG F(ab')2 Secondary Antibody, Rhodamine, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-73293
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Golden Syrian Hamster samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| Rhodamine | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Hamster IgG F(ab')2 fragment | |
| 1 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against Hamster IgG F(c). | |
| Golden Syrian Hamster | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction