Learn More
GNAT1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GNAT1.
Brand: Abnova PAB27001.100uL
Description
Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in rods. This gene is also expressed in other cells, and has been implicated in bitter taste transduction in rat taste cells. Mutations in this gene result in autosomal dominant congenital stationary night blindness. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Specifications
| GNAT1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| GNAT1 | |
| CSNBAD3/GBT1/GNATR | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2779 | |
| Human, Mouse | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of GNAT1. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| P11488 | |
| GNAT1 | |
| A synthetic peptide corresponding to human GNAT1. | |
| 100 μL | |
| Primary | |
| GNAT1 polyclonal antibody detects endogenous levels of GNAT1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only