Learn More
GMCL1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GMCL1.
Brand: Abnova PAB23562.100uL
Description
This gene encodes a nuclear envelope protein that appears to be involved in spermatogenesis, either directly or by influencing genes that play a more direct role in the process. This multi-exon locus is the homolog of the mouse and drosophila germ cell-less gene but the human genome also contains a single-exon locus on chromosome 5 that contains an open reading frame capable of encoding a highly-related protein. [provided by RefSeq
Sequence: WMFLQLVPSWNGSLKQLLTETDVWFSKQRKDFEGMAFLETEQGKPFVSVFRHLRLQYIISDLASARIIEQDAVVPSEWLSSVYKQQWFAMLRAEQDSpecifications
| GMCL1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GMCL1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q96IK5 | |
| GMCL1 | |
| Recombinant protein corresponding to amino acids of human GMCL1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| GMCL1 | |
| BTBD13/GCL/GCL1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 64395 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only