Learn More
GM2A, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GM2A.
Brand: Abnova PAB28161.50ug
Description
This gene encodes a small glycolipid transport protein which acts as a substrate specific co-factor for the lysosomal enzyme beta-hexosaminidase A. Beta-hexosaminidase A, together with GM2 ganglioside activator, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Mutations in this gene result in GM2-gangliosidosis type AB or the AB variant of Tay-Sachs disease. [provided by RefSeq
Specifications
| GM2A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GM2A. | |
| In PBS (0.09% sodium azide) | |
| P17900 | |
| GM2A | |
| A synthetic peptide corresponding to 15 amino acids at internal region of human GM2A. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/ml) | |
| GM2A | |
| SAP-3 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 2760 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only