Learn More
GLRA1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GLRA1.
Brand: Abnova PAB19797.100ug
Description
The protein encoded by this gene is a subunit of a pentameric inhibitory glycine receptor. The receptor mediates postsynaptic inhibition in the central nervous system. Defects in this gene are a cause of startle disease (STHE), also known as hereditary hyperekplexia or congenital stiff-person syndrome. Two transcript variants encoding different isoforms have been found for this gene
Specifications
| GLRA1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GLRA1. | |
| In serum (0.02% sodium azide) | |
| P23415 | |
| GLRA1 | |
| A synthetic peptide corresponding to 16 amino acids near N-terminus of human GLRA1. | |
| RUO | |
| 2741 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:16000-1:60000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user. | |
| GLRA1 | |
| MGC138878/MGC138879/STHE | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only