Learn More
GLE1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GLE1.
Brand: Abnova PAB16679.100ug
Description
This gene encodes a predicted 75kDa polypeptide with high sequence and structure homology to yeast Gle1p, which is nuclear protein with a leucine-rich nuclear export sequence essential for poly(A)+RNA export. Inhibition of human GLE1L by microinjection of antibodies against GLE1L in HeLa cells resulted in inhibition of poly(A)+RNA export. Immunoflourescence studies show that GLE1L is localized at the nuclear pore complexes. This localization suggests that GLE1L may act at a terminal step in the export of mature RNA messages to the cytoplasm. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
| GLE1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GLE1. | |
| In PBS (0.02% sodium azide) | |
| NP_001003722 | |
| GLE1 | |
| A synthetic peptide corresponding to C-terminus 15 amino acids of human GLE1. | |
| RUO | |
| 2733 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| GLE1 | |
| GLE1L/LCCS/LCCS1/hGLE1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only