Learn More
GLDC, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GLDC.
Brand: Abnova PAB21084.100uL
Description
The enzyme system for cleavage of glycine (glycine cleavage system; GCS; EC 2.1.2.10), which is confined to the mitochondria, is composed of 4 protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme), and L protein (a lipoamide dehydrogenase). Glycine encephalopathy (GCE; MIM 605899) may be due to a defect in any one of these enzymes; see MIM 238310, MIM 238330, and MIM 238331.[supplied by OMIM
Sequence: ILNANYMAKRLETHYRILFRGARGYVGHEFILDTRPFKKSANIEAVDVAKRLQDYGFHAPTMSWPVAGTLMVEPTESEDKAELDRFCDAMISIRQEIADIEEGRIDPRVNPLKMSPHSLTCVTSSHWDRPYSREVAAFPLPFVKPENKSpecifications
| GLDC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GLDC. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P23378 | |
| GLDC | |
| Recombinant protein corresponding to amino acids of human GLDC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| GLDC | |
| GCE/GCSP/HYGN1/MGC138198/MGC138200/NKH | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2731 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only