Learn More
GJC3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant GJC3.
Brand: Abnova PAB20786.100uL
Description
Connexins, such as GJC3, are involved in the formation of gap junctions, intercellular conduits that directly connect the cytoplasms of contacting cells. Each gap junction channel is formed by docking of 2 hemichannels, each of which contains 6 connexin subunits (Sohl et al., 2003 [PubMed 12881038]).[supplied by OMIM
Sequence: RTWKHKSSSSKYFLTSESTRRHKKATDSLPVVETKEQFQEAVPGRSLAQEKQRPVGPRDASpecifications
| GJC3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant GJC3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8NFK1 | |
| GJC3 | |
| Recombinant protein corresponding to amino acids of human GJC3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| GJC3 | |
| CX30.2/CX31.3/Cx29/GJE1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 349149 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only