Learn More
GJC1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GJC1.
Brand: Abnova PAB3593.400uL
Description
This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. Alternatively spliced transcript variants encoding the same isoform have been described. [provided by RefSeq
Specifications
| GJC1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GJC1. | |
| In PBS (0.09% sodium azide) | |
| P36383 | |
| GJC1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human GJC1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| GJC1 | |
| CX45/DKFZp686P0738/GJA7 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 10052 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only