Learn More
GJA1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GJA1.
Brand: Abnova PAB24856.100uL
Description
This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. The encoded protein is the major protein of gap junctions in the heart that are thought to have a crucial role in the synchronized contraction of the heart and in embryonic development. A related intronless pseudogene has been mapped to chromosome 5. Mutations in this gene have been associated with oculodentodigital dysplasia and heart malformations. [provided by RefSeq
Specifications
| GJA1 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| GJA1 | |
| GJA1 | |
| A synthetic peptide corresponding to GJA1. | |
| 100 μL | |
| Primary | |
| GJA1 polyclonal antibody detects endogenous levels of GJA1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of GJA1. | |
| In 1X PBS, pH 7.2 (0.05% sodium azide) | |
| CX43/DFNB38/GJAL/ODDD | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2697 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only