missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIT1 (phospho Y554), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GIT1.
Brand: Abnova PAB8161.400uL
This item is not returnable.
View return policy
Specifications
| GIT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of GIT1. | |
| In PBS (0.09% sodium azide) | |
| NP_054749;Q9Y2X7 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 28964 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA | |
| Unconjugated | |
| ELISA (1:1000) Dot Blot (1:500) The optimal working dilution should be determined by the end user. | |
| GIT1 | |
| GIT1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y554 of human GIT1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction