missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gfra3 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Gfra3.
Brand: Abnova PAB0695.100uL
This item is not returnable.
View return policy
Description
Sequence: MGLSWSPRPPLSpecifications
| Gfra3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Gfra3. | |
| Lyophilized from PBS | |
| O35118 | |
| Gfra3 | |
| A synthetic peptide (conjugated with KLH) corresponding to amino acids 1-11 of mouse Gfra3. | |
| RUO | |
| 14587 | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Western Blot (1:500-1:1000) The optimal dilution should be determined by the end user. | |
| Gfra3 | |
| Y15110 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction