Learn More
GFRA2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GFRA2.
Brand: Abnova PAB9890.100ug
Description
Glial cell line-derived neurotrophic factor (GDNF) and neurturin (NTN) are two structurally related, potent neurotrophic factors that play key roles in the control of neuron survival and differentiation. The protein encoded by this gene is a member of the GDNF receptor family. It is a glycosylphosphatidylinositol(GPI)-linked cell surface receptor for both GDNF and NTN, and mediates activation of the RET tyrosine kinase receptor. This encoded protein acts preferentially as a receptor for NTN compared to its other family member, GDNF family receptor alpha 1. This gene is a candidate gene for RET-associated diseases. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| GFRA2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GFRA2. | |
| In PBS (0.02% sodium azide) | |
| GDNFRB/NRTNR-ALPHA/NTNRA/RETL2/TRNR2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.25-0.5 ug/lmL) The optimal working dilution should be determined by the end user. | |
| GFRA2 | |
| GFRA2 | |
| A synthetic peptide corresponding to amino acids 377-391 of human GFRA2. | |
| RUO | |
| 2675 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only