Learn More
GDF9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GDF9.
Brand: Abnova PAB3888.400uL
Description
Growth factors synthesized by ovarian somatic cells directly affect oocyte growth and function. Growth differentiation factor-9 (GDF9) is expressed in oocytes and is thought to be required for ovarian folliculogenesis. GDF9 is a member of the transforming growth factor-beta superfamily. [provided by RefSeq
Specifications
| GDF9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GDF9. | |
| In PBS (0.09% sodium azide) | |
| NP_005251 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 2661 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| GDF9 | |
| GDF9 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human GDF9. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only