Learn More
GDF9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GDF9.
Brand: Abnova PAB25078.100uL
Description
Growth factors synthesized by ovarian somatic cells directly affect oocyte growth and function. Growth differentiation factor-9 (GDF9) is expressed in oocytes and is thought to be required for ovarian folliculogenesis. GDF9 is a member of the transforming growth factor-beta superfamily. [provided by RefSeq
Specifications
| GDF9 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| GDF9 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 2661 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of GDF9. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| GDF9 | |
| A synthetic peptide corresponding to GDF9. | |
| 100 μL | |
| Primary | |
| GDF9 polyclonal antibody detects endogenous levels of GDF9 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only