Learn More
GCS1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GCS1.
Brand: Abnova PAB4280.400uL
Description
This gene encodes the first enzyme in the N-linked oligosaccharide processing pathway. The enzyme cleaves the distal alpha-1,2-linked glucose residue from the Glc(3)-Man(9)-GlcNAc(2) oligosaccharide precursor. This protein is located in the lumen of the endoplasmic reticulum. Defects in this gene are a cause of type IIb congenital disorder of glycosylation (CDGIIb). Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| GCS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GCS1. | |
| In PBS (0.09% sodium azide) | |
| GCS1_HUMAN | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 7841 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| GCS1 | |
| GCS1 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human GCS1. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only